Assay Details
Gene Symbol: Mapk10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270556.1 | NP_001257485.1 | 5 - 6 | 869 | 123 | |
NM_012806.2 | NP_036938.2 | 5 - 6 | 896 | 123 | ||
XM_008769992.1 | XP_008768214.1 | 4 - 5 | 3688 | 123 | ||
GenBank mRNA | DQ377224.1 | - | 2 - 3 | 243 | 123 | |
FQ211777.1 | - | 5 - 6 | 868 | 123 | ||
L27128.1 | - | 5 - 6 | 606 | 123 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
