Assay Details
Gene Symbol: Ephx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034090.3 | NP_001029262.1 | 5 - 6 | 867 | 59 | |
NM_012844.3 | NP_036976.2 | 5 - 6 | 896 | 59 | ||
GenBank mRNA | BC061568.1 | - | 5 - 6 | 827 | 59 | |
M26125.1 | - | 872 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
