Assay Details
Gene Symbol: Hsd17b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012851.2 | NP_036983.1 | 1 - 2 | 141 | 64 | |
GenBank mRNA | BC086365.1 | - | 1 - 2 | 126 | 64 | |
X78811.1 | - | 1 - 2 | 102 | 64 | ||
X97754.1 | - | 1 - 2 | 142 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
