Assay Details
Gene Symbol: Lipe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012859.1 | NP_036991.1 | 7 - 8 | 2549 | 74 | |
XM_006228391.2 | XP_006228453.1 | 7 - 8 | 1682 | 74 | ||
XM_006228393.2 | XP_006228455.1 | 7 - 8 | 1933 | 74 | ||
XM_006228394.2 | XP_006228456.1 | 7 - 8 | 3260 | 74 | ||
XM_008758895.1 | XP_008757117.1 | 7 - 8 | 1666 | 74 | ||
XM_008758896.1 | XP_008757118.1 | 7 - 8 | 1826 | 74 | ||
GenBank mRNA | U40001.1 | - | 7 - 8 | 2549 | 74 | |
X51415.1 | - | 6 - 7 | 2071 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
