Assay Details
Gene Symbol: P4hb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012998.1 | NP_037130.1 | 8 - 9 | 1226 | 82 | |
GenBank mRNA | BC061857.1 | - | 8 - 9 | 1240 | 82 | |
M21018.1 | - | 8 - 9 | 1104 | 82 | ||
X02918.1 | - | 8 - 9 | 1226 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
