Assay Details
Gene Symbol: Rab4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013019.2 | NP_037151.2 | 6 - 7 | 665 | 89 | |
XM_008772599.1 | XP_008770821.1 | 6 - 7 | 1504 | 89 | ||
XM_008772600.1 | XP_008770822.1 | 6 - 7 | 678 | 89 | ||
GenBank mRNA | BC062016.1 | - | 6 - 7 | 665 | 89 | |
X06890.1 | - | 6 - 7 | 597 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
