Assay Details
Gene Symbol: Hspa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013083.2 | NP_037215.1 | 3 - 4 | 533 | 83 | |
GenBank mRNA | BC062017.1 | - | 3 - 4 | 493 | 83 | |
CB578015.1 | - | 3 - 4 | 533 | 83 | ||
M14050.1 | - | 2 - 3 | 409 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
