Assay Details
Gene Symbol: Fh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017005.2 | NP_058701.2 | 7 - 8 | 1110 | 82 | |
GenBank mRNA | BC087598.1 | - | 7 - 8 | 1110 | 82 | |
FQ218764.1 | - | 3 - 4 | 421 | 82 | ||
FQ230216.1 | - | 3 - 4 | 455 | 82 | ||
FQ232207.1 | - | 3 - 4 | 455 | 82 | ||
J04473.1 | - | 7 - 8 | 1121 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
