Assay Details
Gene Symbol: Grn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145842.1 | NP_001139314.1 | 8 - 9 | 884 | 73 | |
NM_017113.2 | NP_058809.2 | 8 - 9 | 884 | 73 | ||
XM_008767981.1 | XP_008766203.1 | 8 - 9 | 965 | 73 | ||
GenBank mRNA | BC072469.1 | - | 8 - 9 | 878 | 73 | |
M97750.1 | - | 7 - 8 | 839 | 73 | ||
X62322.1 | - | 8 - 9 | 858 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
