Assay Details
Gene Symbol: Slc7a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017217.1 | NP_058913.1 | 10 - 11 | 1745 | 67 | |
XM_006257059.2 | XP_006257121.1 | 11 - 12 | 2075 | 67 | ||
GenBank mRNA | AB000113.1 | - | 10 - 11 | 1745 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
