Assay Details
Gene Symbol: Atn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | U31777.1 | - | 3 - 4 | 492 | 81 |
Gene Symbol: LOC100911672 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750658.3 | XP_003750706.1 | 3 - 4 | 479 | 81 | |
XM_006244637.2 | XP_006244699.1 | 3 - 4 | 465 | 81 | ||
XM_008766913.1 | XP_008765135.1 | 3 - 4 | 447 | 81 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
