Assay Details
Gene Symbol: Slc23a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017315.2 | NP_059011.1 | 11 - 12 | 1332 | 89 | |
XM_006254602.2 | XP_006254664.1 | 13 - 14 | 1713 | 89 | ||
GenBank mRNA | AF080452.1 | - | 11 - 12 | 1332 | 89 | |
BC078851.1 | - | 11 - 12 | 1332 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
