Assay Details
Gene Symbol: Srsf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195505.1 | NP_001182434.1 | 704 | 143 | ||
NM_001195506.1 | NP_001182435.1 | 575 | 143 | |||
NM_019257.3 | NP_062130.2 | 579 | 143 | |||
XM_008764878.1 | XP_008763100.1 | 6 - 7 | 755 | 143 | ||
XM_008764879.1 | XP_008763101.1 | 6 - 7 | 626 | 143 | ||
GenBank mRNA | AF020683.1 | - | 5 - 6 | 1876 | 143 | |
BC058479.1 | - | 573 | 143 | |||
L13635.1 | - | 675 | 143 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
