Assay Details
Gene Symbol: Rgs8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019344.2 | NP_062217.1 | 2 - 3 | 305 | 91 | |
XM_006250016.2 | XP_006250078.1 | 2 - 3 | 120 | 91 | ||
XM_006250017.2 | XP_006250079.1 | 2 - 3 | 470 | 91 | ||
XM_008769675.1 | XP_008767897.1 | 2 - 3 | 471 | 91 | ||
XM_008769676.1 | XP_008767898.1 | 2 - 3 | 636 | 91 | ||
GenBank mRNA | AB006013.1 | - | 1 - 2 | 214 | 91 | |
BC089064.1 | - | 2 - 3 | 305 | 91 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AB006013.1, BC089064.1, NM_019344.2, XM_006250016.2, XM_006250017.2, XM_008769675.1, XM_008769676.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
