Assay Details
Gene Symbol: Ppt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019367.2 | NP_062240.1 | 1 - 2 | 129 | 58 | |
XM_008772762.1 | XP_008770984.1 | 1 | 207 | |||
GenBank mRNA | AF061971.1 | - | 1 - 2 | 61 | 58 | |
BC062023.1 | - | 1 - 2 | 91 | 58 | ||
CB736242.1 | - | 1 - 2 | 184 | 58 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AF061971.1, BC062023.1, NM_019367.2, XM_008772762.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
