Assay Details
Gene Symbol: Nptn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019380.1 | NP_062253.1 | 1 - 2 | 236 | 87 | |
XM_006243172.2 | XP_006243234.1 | 1 - 2 | 249 | 87 | ||
GenBank mRNA | X99338.1 | - | 1 - 2 | 236 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
