Assay Details
Gene Symbol: Slc24a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020090.1 | NP_064475.1 | 4 - 5 | 2040 | 138 | |
GenBank mRNA | AF176688.1 | - | 4 - 5 | 2040 | 138 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
