Assay Details
Gene Symbol: Ppp2r2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022209.3 | NP_071545.2 | 4 - 5 | 653 | 66 | |
NR_073588.1 | 7 - 8 | 862 | 66 | |||
GenBank mRNA | AY251277.1 | - | 4 - 5 | 605 | 66 | |
BC078834.1 | - | 7 - 8 | 862 | 66 | ||
D14421.1 | - | 4 - 5 | 428 | 66 | ||
D38260.1 | - | 4 - 5 | 448 | 66 | ||
FQ212581.1 | - | 2 - 3 | 173 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
