Assay Details
Gene Symbol: Acvrl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022441.2 | NP_071886.1 | 5 - 6 | 768 | 60 | |
XM_006242303.2 | XP_006242365.1 | 6 - 7 | 1169 | 60 | ||
XM_006242304.2 | XP_006242366.1 | 6 - 7 | 1773 | 60 | ||
XM_006242305.2 | XP_006242367.1 | 5 - 6 | 1788 | 60 | ||
XM_006242307.2 | XP_006242369.1 | 5 - 6 | 1658 | 60 | ||
XM_008765673.1 | XP_008763895.1 | 6 - 7 | 1832 | 60 | ||
GenBank mRNA | BC083173.1 | - | 5 - 6 | 768 | 60 | |
FQ223580.1 | - | 5 - 6 | 810 | 60 | ||
L36088.1 | - | 6 - 7 | 1183 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
