Assay Details
Gene Symbol: Gale | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080783.2 | NP_542961.2 | 2 - 3 | 155 | 63 | |
XM_006239134.2 | XP_006239196.1 | 2 - 3 | 1120 | 63 | ||
XM_008764197.1 | XP_008762419.1 | 2 - 3 | 386 | 63 | ||
GenBank mRNA | BC097293.1 | - | 2 - 3 | 155 | 63 | |
X53949.1 | - | 1 - 2 | 136 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
