Assay Details
Gene Symbol: Abcg8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130414.2 | NP_569098.2 | 7 - 8 | 1239 | 69 | |
XM_008764453.1 | XP_008762675.1 | 5 - 6 | 1280 | 69 | ||
GenBank mRNA | AF351785.2 | - | 7 - 8 | 1239 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
