Assay Details
Gene Symbol: Olr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133306.2 | NP_579840.2 | 3 - 4 | 512 | 86 | |
GenBank mRNA | AB005900.1 | - | 3 - 4 | 512 | 86 | |
BC097290.1 | - | 3 - 4 | 465 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
