Assay Details
Gene Symbol: Scpep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133383.1 | NP_596874.1 | 9 - 10 | 901 | 77 | |
GenBank mRNA | AF330051.1 | - | 9 - 10 | 901 | 77 | |
FQ235041.1 | - | 1 - 2 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
