Assay Details
Gene Symbol: Hadhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006239786.1 | XP_006239848.1 | 6 - 7 | 838 | 143 | |
XM_006239788.1 | XP_006239850.1 | 6 - 7 | 494 | 143 | ||
XM_006239789.2 | XP_006239851.1 | 6 - 7 | 769 | 143 | ||
GenBank mRNA | BC060545.1 | - | 6 - 7 | 421 | 143 | |
D16479.1 | - | 5 - 6 | 385 | 143 | ||
FM050540.1 | - | 6 - 7 | 449 | 143 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
