Assay Details
Gene Symbol: Hadhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006239786.1 | XP_006239848.1 | 9 - 10 | 1292 | 109 | |
XM_006239788.1 | XP_006239850.1 | 9 - 10 | 948 | 109 | ||
XM_006239789.2 | XP_006239851.1 | 9 - 10 | 1223 | 109 | ||
GenBank mRNA | BC060545.1 | - | 9 - 10 | 875 | 109 | |
D16479.1 | - | 8 - 9 | 839 | 109 | ||
FM038662.1 | - | 2 - 3 | 359 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
