Assay Details
Gene Symbol: Pdgfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012801.1 | NP_036933.1 | 2 - 3 | 420 | 108 | |
GenBank mRNA | L06238.1 | - | 2 - 3 | 420 | 108 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the L06238.1, NM_012801.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
