Assay Details
Gene Symbol: Fgb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020071.2 | NP_064456.2 | 5 - 6 | 833 | 66 | |
XM_006232524.2 | XP_006232586.1 | 5 - 6 | 851 | 66 | ||
XM_008761078.1 | XP_008759300.1 | 5 - 6 | 790 | 66 | ||
GenBank mRNA | AY321323.1 | - | 829 | 66 | ||
AY325147.1 | - | 829 | 66 | |||
AY325153.1 | - | 829 | 66 | |||
BC088102.1 | - | 5 - 6 | 833 | 66 | ||
M27220.1 | - | 2 - 3 | 245 | 66 | ||
U05675.1 | - | 5 - 6 | 834 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
