Assay Details
Gene Symbol: Chkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017177.1 | NP_058873.1 | 1 - 2 | 503 | 59 | |
GenBank mRNA | AB006607.1 | - | 1 - 2 | 503 | 59 | |
BC060515.1 | - | 1 - 2 | 479 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
