Assay Details
Gene Symbol: Chkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017177.1 | NP_058873.1 | 9 - 10 | 1308 | 81 | |
XM_006242181.2 | XP_006242243.2 | 9 - 10 | 1974 | 81 | ||
XM_008765677.1 | XP_008763899.1 | 9 - 10 | 1971 | 81 | ||
GenBank mRNA | AB006607.1 | - | 9 - 10 | 1308 | 81 | |
BC060515.1 | - | 9 - 10 | 1284 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
