Assay Details
Gene Symbol: Senp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023989.1 | NP_076479.1 | 12 - 13 | 1258 | 117 | |
XM_008768805.1 | XP_008767027.1 | 12 - 13 | 1404 | 117 | ||
GenBank mRNA | AF260129.1 | - | 12 - 13 | 1258 | 117 | |
FQ210639.1 | - | 12 - 13 | 1353 | 117 | ||
FQ212134.1 | - | 12 - 13 | 1353 | 117 | ||
FQ222500.1 | - | 11 - 12 | 1201 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
