Assay Details
Gene Symbol: Senp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023989.1 | NP_076479.1 | 7 - 8 | 736 | 137 | |
XM_008768805.1 | XP_008767027.1 | 7 - 8 | 867 | 137 | ||
GenBank mRNA | AF260129.1 | - | 7 - 8 | 736 | 137 | |
FQ212134.1 | - | 7 - 8 | 831 | 137 | ||
FQ222500.1 | - | 6 - 7 | 679 | 137 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
