Assay Details
Gene Symbol: Vdac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031354.1 | NP_112644.1 | 1 - 2 | 145 | 114 | |
XM_006251661.1 | XP_006251723.1 | 2 - 3 | 266 | 114 | ||
GenBank mRNA | AB039663.1 | - | 1 - 2 | 145 | 114 | |
BC063164.1 | - | 1 - 2 | 114 | 114 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AB039663.1, BC063164.1, NM_031354.1, XM_006251661.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
