Assay Details
Gene Symbol: Cd276 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182824.2 | NP_877976.1 | 6 - 7 | 1022 | 117 | |
GenBank mRNA | AY190319.1 | - | 6 - 7 | 1025 | 117 | |
BC100064.1 | - | 6 - 7 | 1022 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
