Assay Details
Gene Symbol: Adprh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183325.2 | NP_899154.2 | 2 - 3 | 459 | 69 | |
XM_006248360.2 | XP_006248422.1 | 2 - 3 | 675 | 69 | ||
GenBank mRNA | BC082065.1 | - | 2 - 3 | 459 | 69 | |
M86341.1 | - | 1 - 2 | 314 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
