Assay Details
Gene Symbol: Gabrg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183327.1 | NP_899156.1 | 7 - 8 | 919 | 65 | |
XM_006246125.2 | XP_006246187.1 | 7 - 8 | 1482 | 65 | ||
GenBank mRNA | AY574252.1 | - | 7 - 8 | 919 | 65 | |
BC092617.1 | - | 2 - 3 | 182 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
