Assay Details
Gene Symbol: Srsf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035255.1 | NP_001030332.1 | 4 - 5 | 618 | 102 | |
XM_006233540.2 | XP_006233602.1 | 5 - 6 | 658 | 102 | ||
XM_006233541.2 | XP_006233603.1 | 5 - 6 | 657 | 102 | ||
XM_006233542.2 | XP_006233604.1 | 5 - 6 | 658 | 102 | ||
XM_006233543.2 | XP_006233605.1 | 6 - 7 | 715 | 102 | ||
XM_008761567.1 | XP_008759789.1 | 5 - 6 | 659 | 102 | ||
XM_008761568.1 | XP_008759790.1 | 6 - 7 | 3706 | 102 | ||
XM_008761569.1 | XP_008759791.1 | 6 - 7 | 722 | 102 | ||
XM_008761570.1 | XP_008759792.1 | 2 - 3 | 906 | 102 | ||
GenBank mRNA | BC099157.1 | - | 4 - 5 | 618 | 102 | |
FQ209707.1 | - | 5 - 6 | 624 | 102 | ||
FQ234514.1 | - | 4 - 5 | 633 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
