Assay Details
Gene Symbol: Eno1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109908.1 | NP_001103378.1 | 4 - 5 | 404 | 91 | |
NM_012554.3 | NP_036686.2 | 4 - 5 | 355 | 91 | ||
GenBank mRNA | BC063174.1 | - | 4 - 5 | 348 | 91 | |
BC078896.1 | - | 4 - 5 | 404 | 91 | ||
BC081847.1 | - | 4 - 5 | 328 | 91 | ||
BC090069.1 | - | 4 - 5 | 285 | 91 | ||
BC091572.1 | - | 4 - 5 | 325 | 91 | ||
BC097343.1 | - | 4 - 5 | 321 | 91 | ||
X02610.1 | - | 4 - 5 | 344 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
