Assay Details
Gene Symbol: Cyp51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012941.2 | NP_037073.1 | 7 - 8 | 1131 | 145 | |
GenBank mRNA | AY724494.1 | - | 7 - 8 | 1453 | 145 | |
BC087033.1 | - | 7 - 8 | 1131 | 145 | ||
D55681.1 | - | 7 - 8 | 1184 | 145 | ||
U17697.1 | - | 7 - 8 | 1297 | 145 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
