Assay Details
Gene Symbol: Lypla1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013006.1 | NP_037138.1 | 1 - 2 | 145 | 75 | |
GenBank mRNA | BC085750.1 | - | 1 - 2 | 83 | 75 | |
D63885.1 | - | 1 - 2 | 145 | 75 | ||
U97146.1 | - | 1 - 2 | 173 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
