Assay Details
Gene Symbol: Got2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013177.2 | NP_037309.1 | 4 - 5 | 555 | 73 | |
GenBank mRNA | BC061792.1 | - | 4 - 5 | 503 | 73 | |
M18467.1 | - | 4 - 5 | 526 | 73 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
