Assay Details
Gene Symbol: Ppp2ca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017039.2 | NP_058735.1 | 1 - 2 | 300 | 81 | |
GenBank mRNA | BC070914.1 | - | 1 - 2 | 309 | 81 | |
BC072531.1 | - | 1 - 2 | 300 | 81 | ||
M33114.1 | - | 1 - 2 | 216 | 81 | ||
X14159.1 | - | 1 - 2 | 216 | 81 | ||
X16043.1 | - | 1 - 2 | 275 | 81 |
Gene Symbol: LOC103694903 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773894.1 | XP_008772116.1 | 795 | 81 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
