Assay Details
Gene Symbol: Gpx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017165.2 | NP_058861.3 | 1 - 2 | 154 | 102 | |
GenBank mRNA | CB582508.1 | - | 1 - 2 | 178 | 102 | |
FQ231108.1 | - | 1 - 2 | 120 | 102 | ||
U37427.1 | - | 1 - 2 | 148 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
