Assay Details
Gene Symbol: Slc25a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017307.3 | NP_059003.1 | 1 - 2 | 213 | 86 | |
GenBank mRNA | BC100077.1 | - | 1 - 2 | 197 | 86 | |
CB758644.1 | - | 1 - 2 | 229 | 86 | ||
FQ214871.1 | - | 1 - 2 | 203 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
