Assay Details
Gene Symbol: Matr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019149.3 | NP_062022.2 | 10 - 11 | 1981 | 150 | |
XM_006254534.2 | XP_006254596.1 | 10 - 11 | 2003 | 150 | ||
XM_006254535.2 | XP_006254597.1 | 10 - 11 | 1954 | 150 | ||
GenBank mRNA | AB205483.1 | - | 9 - 10 | 2751 | 150 | |
M63485.1 | - | 10 - 11 | 1966 | 150 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
