Assay Details
Gene Symbol: Matr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019149.3 | NP_062022.2 | 3 - 4 | 1215 | 65 | |
XM_006254534.2 | XP_006254596.1 | 3 - 4 | 1237 | 65 | ||
XM_006254535.2 | XP_006254597.1 | 3 - 4 | 1188 | 65 | ||
GenBank mRNA | AB205483.1 | - | 2 - 3 | 1985 | 65 | |
BC062231.1 | - | 3 - 4 | 1238 | 65 | ||
BC168723.1 | - | 3 - 4 | 1187 | 65 | ||
M63485.1 | - | 3 - 4 | 1200 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
