Assay Details
Gene Symbol: Crip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022501.1 | NP_071946.1 | 1 - 2 | 95 | 82 | |
GenBank mRNA | BC061774.1 | - | 1 - 2 | 64 | 82 | |
D17512.1 | - | 1 - 2 | 95 | 82 | ||
FQ226314.1 | - | 1 - 2 | 103 | 82 | ||
FQ227529.1 | - | 1 - 2 | 105 | 82 | ||
FQ230331.1 | - | 1 - 2 | 105 | 82 | ||
FQ233584.1 | - | 1 - 2 | 109 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
