Assay Details
Gene Symbol: Ptbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271057.1 | NP_001257986.1 | 4 - 5 | 495 | 88 | |
NM_022516.5 | NP_071961.2 | 4 - 5 | 495 | 88 | ||
GenBank mRNA | BC061858.1 | - | 4 - 5 | 490 | 88 | |
CK365398.1 | - | 1 - 2 | 81 | 88 | ||
FN803753.1 | - | 4 - 5 | 465 | 88 | ||
X60789.1 | - | 4 - 5 | 458 | 88 | ||
X74565.1 | - | 4 - 5 | 476 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
