Assay Details
Gene Symbol: Ube2g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022690.2 | NP_073181.1 | 5 - 6 | 855 | 146 | |
GenBank mRNA | AF099093.1 | - | 5 - 6 | 822 | 150 | |
BC086980.1 | - | 5 - 6 | 855 | 146 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
ubiquitin-dependent protein catabolic process
protein ubiquitination
cellular protein catabolic process
protein K63-linked ubiquitination
protein K48-linked ubiquitination
ubiquitin-protein transferase activity
ATP binding
ligase activity
ubiquitin-like protein transferase activity
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity
protein polyubiquitination
ubiquitin-dependent protein catabolic process
protein ubiquitination
cellular protein catabolic process
protein K63-linked ubiquitination
protein K48-linked ubiquitination
ubiquitin-protein transferase activity
ATP binding
ligase activity
ubiquitin-like protein transferase activity
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P000469 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878417 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891856 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468301 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526574 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260459 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409452 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906250 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961382 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769960 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886336 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915587 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159632 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636026 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360556 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58351 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35187 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519931 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734899 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 549335 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488853 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521840 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487278 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598158 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602586 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715993 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760963 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913031 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298396 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693351 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640590 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345786 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270469 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767384 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415918 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489739 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777818 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826051 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27366 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561988 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760705 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40674 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248376 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482856 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695169 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140419 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396625 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469306 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831242 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391728 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443976 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76676 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965367 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285545 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538027 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539389 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272183 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250416 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663479 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92333 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 42511 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37333 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65826 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50237 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48352 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6591 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4391 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53101 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84953 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P000469 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P000469 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P000469 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P247667 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701259524 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700641231 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 27246 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 3184 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 100898 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 68029 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 11020 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700641231 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 701259524 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10735489 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10735490 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10735484 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10735486 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10735491 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10735488 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10735487 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10735485 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10735483 | - | - |
Rat Genome 230 2.0 Array | 1398864_PM_at | - | - |
Rat Genome 230 2.0 Array | 1370501_at | - | - |
Rat Genome 230 2.0 Array | 1398864_at | - | - |
Rat Genome 230 2.0 Array | 1370501_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1398864_at | - | - |
Affymetrix HT Rat Focus Array | 1370501_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1370501_at | - | - |
Affymetrix HT Rat Focus Array | 1398864_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1000009067.rn.1 | TC1000001008.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000009066.rn.1 | TC1000001008.rn.1 | RTA TC |
Rat Expression Set 230 | 1398864_PM_at | - | - |
Rat Expression Set 230 | 1398864_at | - | - |
Rat Expression Set 230 | 1370501_PM_at | - | - |
Rat Expression Set 230 | 1370501_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10735483 | - | - |
Rat Genome U34 Set | AF099093_g_at | - | - |
Rat Genome U34 Set | AF099093_at | - | - |
Rat Genome U34 Set | rc_AA851376_at | - | - |
Rat Genome U34 Set | rc_AA996896_at | - | - |
Rat Genome U34 Set | rc_AI229984_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21782218 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1360689 | - | - |
RatRef-12 Expression BeadChip | ILMN_63582 | - | - |
