Assay Details
Gene Symbol: Prmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024363.1 | NP_077339.1 | 1 - 2 | 34 | 62 | |
GenBank mRNA | BC078815.1 | - | 1 - 2 | 39 | 62 | |
FQ213198.1 | - | 1 - 2 | 47 | 62 | ||
FQ219843.1 | - | 1 - 2 | 45 | 62 | ||
FQ228625.1 | - | 1 - 2 | 316 | 62 | ||
FQ233897.1 | - | 1 - 2 | 47 | 62 | ||
U60882.1 | - | 1 - 2 | 34 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
