Assay Details
Gene Symbol: LOC686129 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008767400.1 | XP_008765622.1 | 2 | 1585 | 123 |
Gene Symbol: Bmpr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030849.1 | NP_110476.1 | 10 - 11 | 1565 | 123 | |
GenBank mRNA | D17667.1 | - | 9 - 10 | 1340 | 123 | |
D38082.1 | - | 9 - 10 | 1422 | 123 | ||
S75359.1 | - | 10 - 11 | 1565 | 123 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
